siRNA Details
| Gene Symbol: Ptpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012637.2 | NP_036769.1 | 3 | 294 | ||
| XM_006235638.2 | XP_006235700.1 | 3 | 363 | |||
| XM_006235639.2 | XP_006235701.1 | 3 | 364 | |||
| GenBank mRNA | BC081829.1 | - | 3 | 294 | 7135855 | |
| M33962.1 | - | 3 | 336 | |||

Ptpn1
Genomic Map