siRNA Details
| Gene Symbol: Eef1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012660.2 | NP_036792.2 | 4, 5 | 945 | ||
| XM_006235735.2 | XP_006235797.1 | 4, 5 | 763 | |||
| XM_008762467.1 | XP_008760689.1 | 4, 5 | 717 | |||
| GenBank mRNA | BC074016.1 | - | 4, 5 | 945 | 7105980 | |
| M62751.1 | - | 4, 5 | 732 | |||

Eef1a2
Genomic Map