siRNA Details
| Gene Symbol: Vamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012663.2 | NP_036795.1 | 3 | 261 | ||
| XM_006246593.2 | XP_006246655.1 | 3 | 679 | |||
| GenBank mRNA | AJ133104.1 | - | 3 | 245 | ||
| BC074003.1 | - | 3 | 261 | 7097683 | ||
| M24105.1 | - | 2 | 199 | |||

Vamp2
Genomic Map