siRNA Details
Gene Symbol: Hnf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012669.1 | NP_036801.1 | 1 | 223 | ||
GenBank mRNA | J03170.1 | - | 1 | 223 | ||
X54423.1 | - | 1 | 73 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hnf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012669.1 | NP_036801.1 | 1 | 223 | ||
GenBank mRNA | J03170.1 | - | 1 | 223 | ||
X54423.1 | - | 1 | 73 |