siRNA Details
Gene Symbol: Thrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270854.1 | NP_001257783.1 | 5 | 345 | ||
NM_012672.3 | NP_036804.2 | 5 | 345 | |||
XM_008770500.1 | XP_008768722.1 | 3 | 257 | |||
GenBank mRNA | DQ191165.1 | - | 2 | 92 | ||
J03819.1 | - | 5 | 341 | |||
J03933.1 | - | 5 | 342 |
