siRNA Details
| Gene Symbol: Thy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012673.2 | NP_036805.1 | 4 | 675 | ||
| GenBank mRNA | CO561977.1 | - | 1 | 127 | ||
| FQ212850.1 | - | 2 | 324 | |||
| M10666.1 | - | 2 | 525 | |||
| M18002.1 | - | 2 | 525 | |||
| X03150.1 | - | 4 | 626 | |||

Thy1
Genomic Map