siRNA Details
| Gene Symbol: Tpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012678.2 | NP_036810.1 | 3 | 319 | ||
| GenBank mRNA | FQ225054.1 | - | 330 | |||
| FQ225082.1 | - | 330 | ||||
| J02780.1 | - | 3 | 322 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpm4| Gene Symbol: Tpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012678.2 | NP_036810.1 | 3 | 319 | ||
| GenBank mRNA | FQ225054.1 | - | 330 | |||
| FQ225082.1 | - | 330 | ||||
| J02780.1 | - | 3 | 322 | |||