siRNA Details
| Gene Symbol: Tpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012678.2 | NP_036810.1 | 3 | 323 | ||
| GenBank mRNA | J02780.1 | - | 3 | 326 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpm4| Gene Symbol: Tpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012678.2 | NP_036810.1 | 3 | 323 | ||
| GenBank mRNA | J02780.1 | - | 3 | 326 | ||