siRNA Details
| Gene Symbol: Ugt1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012683.2 | NP_036815.1 | 1 | 195 | ||
| GenBank mRNA | AF461736.1 | - | 1 | 198 | ||
| AY435128.1 | - | 1 | 179 | |||
| S70360.1 | - | 1 | 215 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugt1a1| Gene Symbol: Ugt1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012683.2 | NP_036815.1 | 1 | 195 | ||
| GenBank mRNA | AF461736.1 | - | 1 | 198 | ||
| AY435128.1 | - | 1 | 179 | |||
| S70360.1 | - | 1 | 215 | |||