siRNA Details
| Gene Symbol: Vsnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012686.2 | NP_036818.2 | 4 | 773 | ||
| GenBank mRNA | BC092197.1 | - | 4 | 773 | 7313365 | |
| D10666.1 | - | 4 | 781 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vsnl1| Gene Symbol: Vsnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012686.2 | NP_036818.2 | 4 | 773 | ||
| GenBank mRNA | BC092197.1 | - | 4 | 773 | 7313365 | |
| D10666.1 | - | 4 | 781 | |||