siRNA Details
| Gene Symbol: Tbxas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012687.1 | NP_036819.1 | 5 | 491 | ||
| XM_008762759.1 | XP_008760981.1 | 5 | 552 | |||
| XM_008762760.1 | XP_008760982.1 | 3 | 1949 | |||
| XM_008762761.1 | XP_008760983.1 | 5 | 542 | |||
| GenBank mRNA | D28773.1 | - | 5 | 491 | ||
| D31798.1 | - | 5 | 413 | |||

Tbxas1
Genomic Map