siRNA Details
| Gene Symbol: Cckar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012688.3 | NP_036820.1 | 1, 2 | 305 | ||
| XM_008770188.1 | XP_008768410.1 | 2, 3 | 489 | |||
| GenBank mRNA | M88096.1 | - | 1, 2 | 309 | ||
More Information
Set Membership: |
|

Cckar
Genomic Map