siRNA Details
| Gene Symbol: Abcb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012690.2 | NP_036822.1 | 6 | 453 | ||
| XM_006235999.2 | XP_006236061.1 | 4 | 513 | |||
| XM_006236000.1 | XP_006236062.1 | 2 | 176 | |||
| XM_008762697.1 | XP_008760919.1 | 6 | 635 | |||
| GenBank mRNA | L15079.1 | - | 6 | 455 | ||

Abcb4
Genomic Map