siRNA Details
| Gene Symbol: Slc6a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012694.2 | NP_036826.1 | 6, 7 | 1014 | ||
| GenBank mRNA | M80233.1 | - | 6, 7 | 1014 | ||
| M80570.1 | - | 988 | ||||
| S76145.1 | - | 5, 6 | 946 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc6a3| Gene Symbol: Slc6a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012694.2 | NP_036826.1 | 6, 7 | 1014 | ||
| GenBank mRNA | M80233.1 | - | 6, 7 | 1014 | ||
| M80570.1 | - | 988 | ||||
| S76145.1 | - | 5, 6 | 946 | |||