siRNA Details
| Gene Symbol: Stx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012700.2 | NP_036832.1 | 4, 5 | 339 | ||
| XM_008759859.1 | XP_008758081.1 | 4, 5 | 876 | |||
| XM_008759860.1 | XP_008758082.1 | 4, 5 | 277 | |||
| GenBank mRNA | M95735.1 | - | 4, 5 | 498 | ||

Stx1b
Genomic Map