siRNA Details
| Gene Symbol: Slc16a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012716.2 | NP_036848.1 | 2 | 275 | ||
| GenBank mRNA | BC078877.1 | - | 2 | 275 | 7122399 | |
| D63834.1 | - | 289 | ||||
| X86216.1 | - | 295 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc16a1| Gene Symbol: Slc16a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012716.2 | NP_036848.1 | 2 | 275 | ||
| GenBank mRNA | BC078877.1 | - | 2 | 275 | 7122399 | |
| D63834.1 | - | 289 | ||||
| X86216.1 | - | 295 | ||||