siRNA Details
| Gene Symbol: Mobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012720.1 | NP_036852.1 | 4 | 558 | ||
| GenBank mRNA | X89637.1 | - | 4 | 558 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mobp| Gene Symbol: Mobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012720.1 | NP_036852.1 | 4 | 558 | ||
| GenBank mRNA | X89637.1 | - | 4 | 558 | ||