siRNA Details
| Gene Symbol: P2rx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012721.2 | NP_036853.2 | 3 | 457 | ||
| XM_008768851.1 | XP_008767073.1 | 3 | 647 | |||
| XM_008768852.1 | XP_008767074.1 | 3 | 332 | |||
| GenBank mRNA | X92070.1 | - | 3 | 338 | ||
| X97376.1 | - | 3 | 468 | |||

P2rx6
Genomic Map