siRNA Details
| Gene Symbol: Fcer1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012724.4 | NP_036856.1 | 4 | 794 | ||
| XM_008769780.1 | XP_008768002.1 | 3 | 1499 | |||
| XM_008769781.1 | XP_008768003.1 | 3 | 1499 | |||
| GenBank mRNA | J03606.1 | - | 4 | 794 | ||
| M17153.1 | - | 4 | 798 | |||
| M21622.1 | - | 4 | 857 | |||
| M21623.1 | - | 4 | 661 | |||

Fcer1a
Genomic Map