siRNA Details
| Gene Symbol: Camk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012727.3 | NP_036859.2 | 8 | 723 | ||
| XM_006254523.2 | XP_006254585.1 | 7 | 665 | |||
| GenBank mRNA | BC128706.1 | - | 8 | 723 | 7455982 | |
| M63334.1 | - | 617 | ||||
| M64757.1 | - | 6 | 713 | |||
| S65840.1 | - | 7 | 648 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009793, BC070420, AK030219, AK043114, X58995.

Camk4
Genomic Map