siRNA Details
| Gene Symbol: Gpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012736.1 | NP_036868.1 | 9 | 1209 | ||
| GenBank mRNA | BC083565.1 | - | 9 | 1167 | 7134834 | |
| FQ223746.1 | - | 1465 | ||||
| U08027.1 | - | 9 | 1188 | |||
| X78593.1 | - | 9 | 1209 | |||

Gpd2
Genomic Map