siRNA Details
| Gene Symbol: Th | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012740.3 | NP_036872.1 | 5, 6 | 664 | ||
| GenBank mRNA | M10244.1 | - | 5, 6 | 664 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Th| Gene Symbol: Th | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012740.3 | NP_036872.1 | 5, 6 | 664 | ||
| GenBank mRNA | M10244.1 | - | 5, 6 | 664 | ||