siRNA Details
| Gene Symbol: Foxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012742.1 | NP_036874.1 | 3 | 1956 | ||
| GenBank mRNA | X55955.1 | - | 3 | 1956 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxa1| Gene Symbol: Foxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012742.1 | NP_036874.1 | 3 | 1956 | ||
| GenBank mRNA | X55955.1 | - | 3 | 1956 | ||