siRNA Details
| Gene Symbol: Stat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012747.2 | NP_036879.1 | 3 | 412 | ||
| XM_006247257.2 | XP_006247319.1 | 3 | 436 | |||
| XM_006247258.2 | XP_006247320.1 | 3 | 436 | |||
| XM_006247259.2 | XP_006247321.1 | 3 | 436 | |||
| GenBank mRNA | BC087025.1 | - | 3 | 412 | 7110655 | |
| X91810.1 | - | 2 | 206 | |||

Stat3
Genomic Map