siRNA Details
| Gene Symbol: Slc2a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012751.1 | NP_036883.1 | 4 | 477 | ||
| XM_006246596.2 | XP_006246658.1 | 5 | 450 | |||
| GenBank mRNA | BC085757.1 | - | 4 | 522 | 7192278 | |
| D28561.1 | - | 4 | 477 | |||
| FQ216450.1 | - | 5 | 416 | |||
| FQ232657.1 | - | 4 | 536 | |||
| FQ233419.1 | - | 4 | 512 | |||
| J04524.1 | - | 4 | 459 | |||
| KJ696745.1 | - | 4 | 362 | |||
| M25482.1 | - | 4 | 442 | |||
| X14771.1 | - | 4 | 490 | |||

Slc2a4
Genomic Map