siRNA Details
| Gene Symbol: Cyp17a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012753.2 | NP_036885.1 | 5 | 840 | ||
| XM_006231434.2 | XP_006231496.1 | 6 | 1163 | |||
| XM_006231435.2 | XP_006231497.1 | 6 | 1132 | |||
| GenBank mRNA | BC078898.1 | - | 5 | 840 | 7111017 | |
| M22204.1 | - | 5 | 842 | |||
| M31681.1 | - | 5 | 852 | |||
| X14086.1 | - | 5 | 842 | |||

Cyp17a1
Genomic Map