siRNA Details
| Gene Symbol: Plagl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012760.3 | NP_036892.1 | 6, 7 | 699 | ||
| GenBank mRNA | U72620.1 | - | 6, 7 | 699 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plagl1| Gene Symbol: Plagl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012760.3 | NP_036892.1 | 6, 7 | 699 | ||
| GenBank mRNA | U72620.1 | - | 6, 7 | 699 | ||