siRNA Details
| Gene Symbol: LPAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278000.1 | NP_001264929.1 | 5 | 1086 | ||
| NM_005296.2 | NP_005287.1 | 2 | 871 | |||
| XM_005262126.3 | XP_005262183.1 | 5 | 1289 | |||
| XM_006724639.3 | XP_006724702.1 | 4 | 1001 | |||
| XM_017029437.1 | XP_016884926.1 | 5 | 2348 | |||
| XM_017029438.1 | XP_016884927.1 | 3 | 1024 | |||
| GenBank mRNA | AK314134.1 | - | 2 | 865 | ||
| BC069996.1 | - | 1 | 527 | 6971739 | ||
| BC074722.2 | - | 1 | 491 | 30915172 | ||
| BC095538.1 | - | 1 | 640 | 30706564 | ||
| U90322.1 | - | 1 | 532 | |||

LPAR4
Genomic Map