siRNA Details
| Gene Symbol: Dyrk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012791.2 | NP_036923.1 | 5 | 510 | ||
| XM_006248031.2 | XP_006248093.1 | 5 | 3455 | |||
| XM_008768562.1 | XP_008766784.1 | 4 | 2718 | |||
| XM_008768563.1 | XP_008766785.1 | 6 | 3591 | |||
| XM_008768564.1 | XP_008766786.1 | 5 | 557 | |||
| XM_008768565.1 | XP_008766787.1 | 5 | 632 | |||
| GenBank mRNA | X79769.1 | - | 5 | 512 | ||

Dyrk1a
Genomic Map