siRNA Details
| Gene Symbol: ITGB1BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012278.2 | NP_036410.1 | 4 | 322 | ||
| GenBank mRNA | AF110225.1 | - | 4 | 276 | ||
| AF140690.1 | - | 4 | 292 | |||
| BC108901.2 | - | 3 | 549 | 40004692 | ||
| DA895605.1 | - | 4 | 325 | |||

ITGB1BP2
Genomic Map