siRNA Details
Gene Symbol: Abcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012804.1 | NP_036936.1 | 6, 7 | 548 | ||
GenBank mRNA | D90038.1 | - | 6, 7 | 548 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Abcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012804.1 | NP_036936.1 | 6, 7 | 548 | ||
GenBank mRNA | D90038.1 | - | 6, 7 | 548 |