siRNA Details
| Gene Symbol: Mapk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270556.1 | NP_001257485.1 | 8 | 1094 | ||
| NM_012806.2 | NP_036938.2 | 8 | 1121 | |||
| XM_008769992.1 | XP_008768214.1 | 7 | 3913 | |||
| GenBank mRNA | DQ377224.1 | - | 5 | 468 | ||
| FQ211777.1 | - | 8 | 1093 | |||
| L27128.1 | - | 8 | 831 | |||

Mapk10
Genomic Map