siRNA Details
| Gene Symbol: Smo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012807.1 | NP_036939.1 | 8 | 1423 | ||
| GenBank mRNA | U84402.1 | - | 8 | 1423 | ||
More Information
Other Species Targeted
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smo| Gene Symbol: Smo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012807.1 | NP_036939.1 | 8 | 1423 | ||
| GenBank mRNA | U84402.1 | - | 8 | 1423 | ||
Set Membership: |
|