siRNA Details
| Gene Symbol: Sycp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012810.1 | NP_036942.1 | 6 | 419 | ||
| GenBank mRNA | X67805.1 | - | 6 | 419 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sycp1| Gene Symbol: Sycp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012810.1 | NP_036942.1 | 6 | 419 | ||
| GenBank mRNA | X67805.1 | - | 6 | 419 | ||