siRNA Details
Gene Symbol: Sycp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012810.1 | NP_036942.1 | 7, 8 | 641 | ||
GenBank mRNA | X67805.1 | - | 7, 8 | 641 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sycp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012810.1 | NP_036942.1 | 7, 8 | 641 | ||
GenBank mRNA | X67805.1 | - | 7, 8 | 641 |