siRNA Details
| Gene Symbol: Alox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012822.1 | NP_036954.1 | 7 | 1044 | ||
| XM_006237140.2 | XP_006237202.1 | 7 | 1081 | |||
| GenBank mRNA | J03960.1 | - | 7 | 1044 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alox5| Gene Symbol: Alox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012822.1 | NP_036954.1 | 7 | 1044 | ||
| XM_006237140.2 | XP_006237202.1 | 7 | 1081 | |||
| GenBank mRNA | J03960.1 | - | 7 | 1044 | ||