siRNA Details
| Gene Symbol: Ankrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013220.1 | NP_037352.1 | 3 | 392 | ||
| XM_008760363.1 | XP_008758585.1 | 4 | 452 | |||
| GenBank mRNA | BC072699.1 | - | 3 | 384 | 7105899 | |
| L81174.1 | - | 3 | 342 | |||
| U50736.1 | - | 3 | 392 | |||

Ankrd1
Genomic Map