siRNA Details
| Gene Symbol: Adra1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016991.2 | NP_058687.2 | 1 | 956 | ||
| GenBank mRNA | BC070920.1 | - | 1 | 956 | 7105608 | |
| M60655.1 | - | 1 | 581 | |||
| X51585.1 | - | 1 | 807 | |||
More Information
Set Membership: |
|

Adra1b
Genomic Map