siRNA Details
| Gene Symbol: Cyp4b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016999.2 | NP_058695.2 | 12 | 1864 | ||
| GenBank mRNA | BC074012.1 | - | 12 | 1874 | 7100197 | |
| M29853.1 | - | 12 | 1864 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp4b1| Gene Symbol: Cyp4b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016999.2 | NP_058695.2 | 12 | 1864 | ||
| GenBank mRNA | BC074012.1 | - | 12 | 1874 | 7100197 | |
| M29853.1 | - | 12 | 1864 | |||