siRNA Details
| Gene Symbol: Epor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017002.2 | NP_058698.2 | 7 | 950 | ||
| XM_006242602.2 | XP_006242664.1 | 7 | 1056 | |||
| XM_006242603.2 | XP_006242665.1 | 7 | 1553 | |||
| GenBank mRNA | BC089810.1 | - | 7 | 950 | 7375416 | |
| D13566.1 | - | 7 | 868 | |||

Epor
Genomic Map