siRNA Details
| Gene Symbol: Erbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017003.2 | NP_058699.2 | 4 | 575 | ||
| GenBank mRNA | AY116182.1 | - | 4 | 575 | ||
| BC061863.1 | - | 4 | 647 | 5597835 | ||
| X03362.1 | - | 4 | 486 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erbb2| Gene Symbol: Erbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017003.2 | NP_058699.2 | 4 | 575 | ||
| GenBank mRNA | AY116182.1 | - | 4 | 575 | ||
| BC061863.1 | - | 4 | 647 | 5597835 | ||
| X03362.1 | - | 4 | 486 | |||