siRNA Details
| Gene Symbol: Gstm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017014.1 | NP_058710.1 | 5 | 374 | ||
| GenBank mRNA | BC063172.1 | - | 5 | 414 | 6920688 | |
| FJ179401.1 | - | 5 | 338 | |||
| FQ211067.1 | - | 5 | 373 | |||
| FQ212742.1 | - | 5 | 367 | |||
| FQ213204.1 | - | 5 | 367 | |||
| FQ213696.1 | - | 5 | 561 | |||
| FQ214055.1 | - | 5 | 473 | |||
| FQ217092.1 | - | 5 | 440 | |||
| FQ218381.1 | - | 5 | 374 | |||
| FQ218789.1 | - | 5 | 373 | |||
| FQ218795.1 | - | 5 | 410 | |||
| FQ229836.1 | - | 5 | 421 | |||
| FQ229941.1 | - | 5 | 401 | |||
| J02810.1 | - | 5 | 374 | |||
| M11719.1 | - | 5 | 375 | |||
| M28241.1 | - | 5 | 434 | |||
| X04229.1 | - | 5 | 374 | |||

Gstm1
Genomic Map