siRNA Details
| Gene Symbol: Gusb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017015.2 | NP_058711.2 | 4 | 655 | ||
| GenBank mRNA | AY321342.1 | - | 800 | |||
| M13962.1 | - | 4 | 625 | |||
| Y00717.1 | - | 4 | 581 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gusb| Gene Symbol: Gusb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017015.2 | NP_058711.2 | 4 | 655 | ||
| GenBank mRNA | AY321342.1 | - | 800 | |||
| M13962.1 | - | 4 | 625 | |||
| Y00717.1 | - | 4 | 581 | |||