siRNA Details
Gene Symbol: Hdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017016.2 | NP_058712.2 | 2 | 243 | ||
GenBank mRNA | BC128724.1 | - | 2 | 243 | 8375507 | |
M29591.1 | - | 2 | 250 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017016.2 | NP_058712.2 | 2 | 243 | ||
GenBank mRNA | BC128724.1 | - | 2 | 243 | 8375507 | |
M29591.1 | - | 2 | 250 |