siRNA Details
| Gene Symbol: Hgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017017.2 | NP_058713.1 | 2 | 289 | ||
| XM_006235979.2 | XP_006236041.1 | 2 | 384 | |||
| GenBank mRNA | D90102.1 | - | 2 | 289 | ||
| X54400.1 | - | 2 | 188 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hgf| Gene Symbol: Hgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017017.2 | NP_058713.1 | 2 | 289 | ||
| XM_006235979.2 | XP_006236041.1 | 2 | 384 | |||
| GenBank mRNA | D90102.1 | - | 2 | 289 | ||
| X54400.1 | - | 2 | 188 | |||