siRNA Details
| Gene Symbol: Kcnj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017023.1 | NP_058719.1 | 2 | 478 | ||
| XM_008766044.1 | XP_008764266.1 | 3 | 609 | |||
| XM_008766045.1 | XP_008764267.1 | 3 | 1299 | |||
| XM_008766046.1 | XP_008764268.1 | 2 | 487 | |||
| XM_008766047.1 | XP_008764269.1 | 2 | 600 | |||
| XM_008766048.1 | XP_008764270.1 | 2 | 1177 | |||
| GenBank mRNA | L29403.1 | - | 2 | 427 | ||
| X72341.1 | - | 2 | 478 | |||

Kcnj1
Genomic Map