siRNA Details
| Gene Symbol: Mpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017027.1 | NP_058723.1 | 3 | 405 | ||
| GenBank mRNA | BC078859.1 | - | 405 | |||
| K03242.1 | - | 3 | 405 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mpz| Gene Symbol: Mpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017027.1 | NP_058723.1 | 3 | 405 | ||
| GenBank mRNA | BC078859.1 | - | 405 | |||
| K03242.1 | - | 3 | 405 | |||