siRNA Details
| Gene Symbol: Nefm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017029.2 | NP_058725.1 | 1 | 516 | ||
| XM_008770736.1 | XP_008768958.1 | 1 | 559 | |||
| GenBank mRNA | M18628.1 | - | 1 | 494 | ||
| Z12152.1 | - | 1 | 519 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nefm| Gene Symbol: Nefm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017029.2 | NP_058725.1 | 1 | 516 | ||
| XM_008770736.1 | XP_008768958.1 | 1 | 559 | |||
| GenBank mRNA | M18628.1 | - | 1 | 494 | ||
| Z12152.1 | - | 1 | 519 | |||