siRNA Details
| Gene Symbol: Pccb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017030.2 | NP_058726.2 | 9 | 922 | ||
| GenBank mRNA | BC078855.1 | - | 9 | 922 | 7108495 | |
| M14634.1 | - | 9 | 923 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pccb| Gene Symbol: Pccb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017030.2 | NP_058726.2 | 9 | 922 | ||
| GenBank mRNA | BC078855.1 | - | 9 | 922 | 7108495 | |
| M14634.1 | - | 9 | 923 | |||