siRNA Details
| Gene Symbol: Pgm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017033.2 | NP_058729.2 | 5 | 759 | ||
| XM_006238442.2 | XP_006238504.1 | 6 | 950 | |||
| XM_008763853.1 | XP_008762075.1 | 5 | 1302 | |||
| GenBank mRNA | BC099807.1 | - | 5 | 759 | 7937943 | |
| BC128703.1 | - | 5 | 764 | |||
| L11694.1 | - | 5 | 742 | |||

Pgm1
Genomic Map